Blotter Paper, IW Tremont
Catalog #: CAIWT-BLOT88
Supplier: IW Tremont
Specifications
- Dimensions:8×8"
- Description:Blotter Paper
- Supplier No.:IWT-BLOT88
Specifications
About this item
Blotter paper with smooth surface. Made from bright white cellulose filter paper. Material is manufactured from carefully selected, fully bleached and chemically free pure cellulose fiber without additives or binder.
Meets standards specified in TAPPI T 205 sp-06.
Customers Who Bought This Also Bought