Order Entry
ContactUsLinkComponent
12 X 75Mm (5Ml) Test Tube
Polystyrene and Polypropylene Test Tubes, 12×75 mm, Globe Scientific
Catalog #: 89209-796
Supplier:  Globe Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
12 X 75Mm (5Ml) Test Tube
Polystyrene and Polypropylene Test Tubes, 12×75 mm, Globe Scientific
Catalog #: 89209-796
Supplier:  Globe Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise

Specifications

  • Diameter ×L:
    12 x 75 mm
  • Volume:
    5 ml
  • Description:
    Test Tube
  • Material:
    Polystyrene
  • Supplier No.:
    110410

Specifications

About this item

These tubes are molded from either premium-grade polystyrene for clarity, or polypropylene for impact strength and stability to common acids, alkalis, and solvents.

Ordering information: Unless otherwise specified, test tubes are supplied without caps.
Customers Who Bought This Also Bought