Unjacketed Filter Reactor Systems, 20 L, Chemglass
Catalog #: 89209-348
Supplier: Chemglass
Small Business Enterprise
Specifications
- Description:UnJacketed Cylindrical Process Filter Reactor System Complete
- Motor Type:Air motor
- Cat. no.:89209-348
- Supplier No.:CG-1958-F821
Specifications
About this item
These unjacketed filter reactors can be used for single or multi-step reactions and filtrations in the same vessel, production and process scale peptide synthesis.
- 150 °C maximum temperature limit for filter base
- Filter base eliminates material transfers and significantly reduces losses
Complete reactor system on our NEW open air, autocenter stand.
Caution: Filter reactors are for vacuum/gravity filtration use only.
Customers Who Bought This Also Bought