SealMate™ SealPlate® Microplate-Sealing System, Excel Scientific
Catalog #: 89209-150
Supplier: Excel Scientific
Specifications
- Optical property:Transparent
- Sterility:Non sterile
- Description:SealPlate® Film Rolls Only, 2/PK, NS
- Material:Polyester
- Thickness:64,8 µm
- Cat. no.:89209-150
- Supplier No.:RSMSP-2
Specifications
About this item
The dispenser facilitates the quick and easy dispensing of film from a roll allowing microplates to be sealed three times faster than using sheeted films. Cost-effective replacement rolls are sold separately.
- Environmentally friendly; no paper liner to remove and discard; utilizes approximately 72% less material than SealPlate sheeted film
- Each sheet is separated by an adhesive-free, perforated, color-coded green band
- The smooth lower surface of the dispenser can be used to seal the film to the plate
- Dispenser protects unused seals from contamination
- Lower support prop prevents dispensing bill from contacting surfaces to prevent contamination
- Replacement rolls can be easily installed by releasing the locking clasps on the dispenser
- Temperature range: –40 to +120 °C
Ideal for use under the hood; the SealMate dispenser minimizes film contamination.
Application: General laboratory use, short-term storage, incubation and ELISA.
Ordering information: The SealMate SealPlate Starter Kit features a re-usable dispenser and two rolls of perforated film.
Customers Who Bought This Also Bought