ContactUsLinkComponent
RNAse Set 6000 Units 200 Preps
Enzymes and Separation Accessories, Omega Bio-Tek
Catalog #: CA89167-382
Supplier:  Omega Bio-Tek
CAS Number:  
Enzymes and Separation Accessories, Omega Bio-Tek
Catalog #: CA89167-382
Supplier:  Omega Bio-Tek
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    RNase-Free DNase Set
  • Size:
    200 Preps (6000 units)
  • Cat. no.:
    CA89167-382
  • Supplier No.:
    E1091-02
Specifications
About this item
Magnetic separation device-A consists of 24 powerful magnetic rods that fit between the wells of commercially-available 96-well microplates
Magnetic separation device-B consists of a series of ten powerful magnetic disks. Each disk attracts the beads in a 1.5 or 2mL tube fitted in an adjacent hole. After the magnetic beads capture DNA or RNA, the beads are pulled to the side of the tube while the buffer is exchanged. Device holds ten 1.5/2mL tubes or one 15mL tube.
RNase-free DNase set provides a rapid and efficient on-column digestion of DNA during RNA purification using E.Z.N.A.® RNA kits. E-Z 96® vacuum manifold is designed for sample preparation with E-Z 96® DNA/RNA plates. The vacuum manifold replaces many steps to help decrease processing time. OB protease and proteinase K are intended for enzyme digestion during DNA and RNA preparation. Ribonuclease A (RNase A) is endonuclease-free for use in plasmid purification procedures for digestion of RNA. X-Gal and IPTG are commonly used for blue/white screening of bacterial colonies to distinguish bacterial or phage clones after transformation experiments.
Customers Who Bought This Also Bought