VWR® Protective Eyewear with Wraparound Lens
Catalog #: 89187-986
Supplier: VWR International
CSA Certified
Product available on GSA Advantage®
Specifications
- Lens Coating:Scratch-resistant/hardcoat
- Lens Tint:Clear
- Frame Color:Clear
Specifications
About this item
VWR® protective eyewear with wraparound lens offers stylish and secure personal protection, with an array of comfort features to assist the wearer throughout extended use.
- Fit comfortably with half-mask respirators
- Impact-resistant, scratch-resistant polycarbonate lens
- Provides 99.9% UV protection
Super lightweight wraparound eyewear for men and women combines a sleek appearance with rugged durability. Eyewear features rubberized temple tips and an integrated nose piece for extra comfort.
Certifications: ANSI Z87.1 certified.
Customers Who Bought This Also Bought