ContactUsLinkComponent
Lens Tissue Paper 4x6cm
Whatman™ Lens Cleaning Tissue, Whatman products (Cytiva)
Catalog #: CA97002-542
Supplier:  Cytiva
Whatman™ Lens Cleaning Tissue, Whatman products (Cytiva)
Catalog #: CA97002-542
Supplier:  Cytiva
Specifications
  • Description:
    Grade 105 lens cleaning tissue, sheet, 100×150 mm
  • Supplier No.:
    2105-841
Specifications
About this item
These cleaning tissues are suitable for use with lenses and any other optical surfaces made from glass, quartz or plastic.
  • Soft surface
  • Strong and highly absorbent
  • Chemically pure tissue free from silicones and other additives
Customers Who Bought This Also Bought