ELGA Cartridges for Water Purification Systems
Catalog #: CA89204-328
Supplier: ELGA CA
Specifications
- Description:Point-of-Use Microfilter, 0.2 µm
- Supplier No.:LC134
Specifications
About this item
By choosing an ELGA water purification system, you are making an investment in quality, reliability and value.
- Protect your lab equipment
- Avoid erroneous data and ensure productivity
- Gain peace of mind
Non-certified ELGA consumables can use low quality materials, poor technology, and prevent your system from delivering the correct water purity to meet your needs.
Ordering information: It is highly recommended to contact your sales representative for additional information to meet your application, budget, and configuration needs.
Customers Who Bought This Also Bought