ELGA Lights for Water Purification Systems, ELGA LabWater
Catalog #: CA89204-286
Supplier: ELGA CA
Specifications
- Description:UV lamp, 254 nm
- Cat. no.:CA89204-286
- Supplier No.:LC105
Specifications
About this item
UV lamps for water purification sysstem.
- Protect your lab equipment
- Avoid erroneous data and ensure productivity
- Gain peace of mind
Non-certified ELGA consumables can use low quality materials, poor technology, and prevent your system from delivering the correct water purity to meet your needs.
Customers Who Bought This Also Bought