Triple Display Certified Thermometers, Thermco
Catalog #: 89137-070
Supplier: Thermco
Specifications
- Resolution:0,1 °C
- Description:Certified at 4 °C, for Refrigerators
- Accuracy:±0.5 °C
- Range:–50...+70 °C
- Supplier No.:ACC895REF
Specifications
About this item
These large digital thermometers simultaneously show the current, minimum, and maximum temperatures and update continuously
The probe is housed in an unbreakable plastic bottle filled with a buffering media that maintains the thermometer reading when the instrument door is opened. The thermometer and the probe bottle have dual magnets to attach to the instrument. Thermometers also feature a flip-open stand, wall mounting key hole, and an AAA alkaline battery.
An audible alarm sounds when the temperature rises above or below a set temperature. The minimum/maximum feature monitors critical temperature for any period of time
Certifications: Each thermometer is certified and traceable to NIST. Each thermometer is serialized.
Customers Who Bought This Also Bought