Specifications
- Resolution:0.01°
- Description:Thermometer with Two Probes
- Accuracy:±0.3 °C
- Range:-58...+158°F / -50...+70°C
Specifications
About this item
Traceable International Standards Digital Thermometer provides the exact readings required for critical requirements, quality control checks, and routine measurements.
- Stainless steel probe construction
- Available with one probe or two probes
- Monitor inlet and outlet simultaneously
- Visual and audible, high and low settable alarms
Two-channel thermometer is available with one probe or two probes
Absolute high-precision thermometer is ideal for measuring temperatures in water baths, cuvets, gas systems, reagents, incubators, pharmaceuticals, petroleum products, foods, wastewater, and when using pH/conductivity standards.
Display shows minimum/maximum and current temperatures. MIN/MAX monitors readings overnight, on weekends, or for any time period. It displays the exact time and date when the MIN/MAX temperature occurred. Alarm provides two visual (LEDs) and two audio alerts when temperature rises above or falls below high and low set points. Unit displays the exact time and date when dual thermometer alarms are triggered. Alarms are programmable in 0.1° increments. Visual and audible alarms signal continuously (for days) even if temperature returns to non-alarm range. First thermometer ever designed to permit user one-button calibration.
Stainless-steel probe supplied with units shows the slightest temperature change. Probe is triple purpose for liquid, air/gas, and semisolids. Accurate readings with tip penetration of 1/3-inch. Probe has a diameter of 1/8-inch, stem length of 6-1/4 inches, overall length of 9 inches and a cable length of 5 feet.
Two probes are useful in monitoring inlets and outlets; tracking two unique tests simultaneously; and measuring temperatures in two locations. Thermometer range is -58 to 158°F and -50 to 70°C with a resolution of 0.01° and accuracy of ±0.3°C. Unit reads in °C and °F.