Reichert Brake-Chek (F)
Catalog #: 89203-926
Supplier: Reichert Technologies
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$881.80 to $2,807.56
Puritan® Standard Cotton Tipped Applicators, Wood Handle, Sterile, Puritan Medical Products
Supplier: Puritan Medical Products
$127.98 to $1,279.44
Potassium chloride saturated aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$76.04 to $889.72
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Sodium bisulfite ≥90% FCC, purified, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$237.49 to $1,424.99