Precleaned VOA Vials, Environmental Express®
Catalog #: 89202-862
Supplier: Environmental Express
Small Business Enterprise
Specifications
- Cap included:Yes
- Cap Type:Open-top
- Capacity:40.00 ml
- Color:Clear
- Description:Level I quality assured/preserved vials with 2% Sodium Thiosulfate 0.2 ml
- Cat. no.:89202-862
- Supplier No.:APC1556
Specifications
About this item
These volatile organic analysis (VOA) vials are made of borosilicate glass. Choose from an open-top cap (0.125" thick PTFE/silicone septa) or closed-top style cap in a variarty of capacities and clear or amber glass.
Preserved vials come pre-filled with common preservative reagents for convenience and safety.
Certifications: All containers meet one of two levels of pre-cleaning. Level III indicates that containers have not received an EPA washing treatment. Level I indicates that each case of containers has been custody-sealed, lot number-labeled for traceability, and a Certificate of Analysis indicating that the containers received full EPA quality assurance treatment.
BodyTextAccessories: The accessory Foam Block Rack holds 72×40 ml vials. Ideal for storage and shipping 40 ml VOA vials.
Customers Who Bought This Also Bought