To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Dimensions:7 x 10 cm (2 3/4 x 3 15/16")
- Thickness:0.38 mm
- Herzberg flow rate:250 sec./100 mL at 10 cm head of water
- Klemm wicking rate test:626 sec./7.5 cm rise
- Weight:185 g/m²
- Cat. no.:CA28298-030
Specifications
About this item
Grade 703 blotting papers are ideal for use as wicks in protein and nucleic acid blotting.
- Made of 100% cotton fiber
- Uniform and smooth
- Produced using ultrapure water
- Contains no additives
Ideal for use as wicks in protein and nucleic acid blotting. Provides a uniform flow of buffer through the gel to the transfer membrane in a blotting sandwich. Also suitable for use in removing gels from glass supports.