VWR® Grade 707 Blotting Paper, Grade 707
Catalog #: CA28298-014
Supplier: VWR International
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Dimensions:7 x 10 cm (23/4 x 315/16")
- Thickness:2.60 mm
- Herzberg flow rate:120 sec./100 mL at 10 cm head of water
- Klemm wicking rate test:96 sec./7.5 cm rise
- Weight:703 g/m2
- Cat. no.:CA28298-014
- Type:Sheet
Specifications
About this item
This superthick paper is used to complete the blotting sandwich.
- Contains no additives
- Made of 100% cotton fiber
- Produced using ultrapure water
- Cleaner and more consistent than paper towels
Smooth, uniform texture assures continuous contact of buffer from the chamber through the gel and transfer membrane to the opposite pole or weight.
Customers Who Bought This Also Bought