Order Entry
ContactUsLinkComponent
Stablcal* Turbidity Standard
Stablcal® Turbidity Standards, 1.0 to 40 NTU, Hach
Catalog #: CA71002-460
Supplier:  Hach
CAS Number:  
Stablcal* Turbidity Standard
Stablcal® Turbidity Standards, 1.0 to 40 NTU, Hach
Catalog #: CA71002-460
Supplier:  Hach
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Stablcal® Turbidity Standard
  • Size:
    1000 ml bottle
  • Storage temperature:
    5 to 25 °C
  • Cat. no.:
    CA71002-460
  • Supplier No.:
    2660153

Specifications

About this item

Stablcal® Turbidity Standards for all turbidimeters.

-Precise, Pre-mixed Formazin Solutions
-Based on Formazin Primary Standards
-Enhanced Stability Saves Time and Labor
-Confidence in results

  • Precise, premixed formazin solutions
  • Based on formazin primary standards
  • Enhanced stability saves time and labor
  • Confidence in results

Stablcal® Turbidity Standards for all turbidimeters.

USEPA accepted for calibration purposes. Stablcal® Stabilized Formazin Turbidity Standards are true formazin dilutions developed for use in any turbidimeter

Customers Who Bought This Also Bought