ContactUsLinkComponent
Goggle Mono Xtr Shld Blu/Clr
KleenGuard™ V90 Shield Safety Goggles with Face Shield, Kimberly-Clark Professional
Catalog #: 89130-975
Supplier:  Kimberly-Clark
Goggle Mono Xtr Shld Blu/Clr
KleenGuard™ V90 Shield Safety Goggles with Face Shield, Kimberly-Clark Professional
Catalog #: 89130-975
Supplier:  Kimberly-Clark
Specifications
  • Description:
    V90 shield protective goggles
  • Cat. no.:
    89130-975
  • Supplier No.:
    18629
Specifications
About this item
The safety goggles feature KleenGuard™ Monogoggle™ XTR goggles integrated with a face shield to provide full face protection from liquids and particulates without the hassle of primary eye protection underneath a face shield.
  • Indirect ventilation provides increased airflow, minimizing condensation and fogging
  • Curved face shield conforms to the shape of the user's face, and is a splash and impact protector
  • Soft, pliable material around the eyes and nose ensure a comfortable fit
  • Goggles can be used over most prescription eyeglasses
  • Shield is detachable
  • Goggles feature a clear, anti-fog lens and blue frame
Protect yourself with the KleenGuard™ V90 Shield Safety Goggles with Face Shield. It's really like having two products in one, since the face shield mask detaches from the splash goggles when needed. Together, they provide eye and full face protection from liquids and particles without the hassle of primary eye protection underneath a face shield mask.
The protective goggles feature an indirect vent that enables free airflow, while safeguarding against liquids entering the eye area. The goggles portion has a soft, pliable frame that conforms to the face for comfort, a cylindrical lens for increased visibility, and indirect ventilation. They can fit over most prescription eyeglasses, and the face shield mask is curved to conform to the shape of face for added protection. The shield includes an adjustable strap, to provide a snug fit.
Certifications: Meets ANSI Z87.1+ standard for impact protection, with a D3 rating for droplet and splash protection.
Customers Who Bought This Also Bought