To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Capacity:50 ml
- Description:3×4" absorbent sheet
- Cat. no.:89170-926
- Supplier No.:10300
Specifications
About this item
Absorbent sheets are the most cost-effective absorbent product that complies with the regulations governing specimen transport.
- Tissue-based
- Impregnated with super-absorbent polymers to absorb liquids that may escape during transport
Sheets can be purchased in a variety of stock sizes or custom cut to meet your specific requirements.