Order Entry
ContactUsLinkComponent
Tubes Test with Flat Head [standard taper] Stopper
KIMBLE® Test Tubes with Flat Head [ST] Stopper, DWK Life Sciences
Catalog #: KT898000-0005
Tubes Test with Flat Head [standard taper] Stopper
KIMBLE® Test Tubes with Flat Head [ST] Stopper, DWK Life Sciences
Catalog #: KT898000-0005

Specifications

  • Capacity:
    5 ml
  • Ø:
    13 mm
  • Height:
    80 mm
  • ST Stopper No.:
    9
  • Supplier No.:
    898000-0005

Specifications

About this item

Reusable test tubes with flat head stoppers.

  • Round bottom
  • Uniform wall thickness for maximum service
  • Solid, flat head standard taper stopper

Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.

Customers Who Bought This Also Bought