ContactUsLinkComponent
Tissue grinders Potter-Elvehjem PTFE Pestle and Glass Tube
Kimble® Potter-Elvehjem Tissue Grinder Components, DWK Life Sciences
Catalog #: KT885512-0023
Tissue grinders Potter-Elvehjem PTFE Pestle and Glass Tube
Kimble® Potter-Elvehjem Tissue Grinder Components, DWK Life Sciences
Catalog #: KT885512-0023
Specifications
  • Description:
    Tube for Potter-Elvehjem Tissue Grinder
  • Size:
    23
  • Cat. no.:
    KT885512-0023
  • Supplier No.:
    885512-0023
Specifications
About this item
A graduated glass tube and a finely machined PTFE pestle comprise this popular style.
  • Graduations are fused into the glass
  • Replacement components are completely interchangeable
Radial serrations on the lower portion of the pestle deliver the homogenate into the cylindrical portion of the tube.
Customers Who Bought This Also Bought