KONTES® Pennyhead [ST] Stoppers, Full Length, Kimble Chase
Catalog #: KT851000-1938
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- ST Joint No.:19/38
- Cat. no.:KT851000-1938
- Supplier No.:851000-1938
Specifications
About this item
Glass [ST] stoppers for use in the same size outer joints
Sizes [ST]10/30 through [ST]19/38 are solid. Sizes [ST]24/40 and larger are hollow.
Customers Who Bought This Also Bought