To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:EcoLite(+)™
- Size:4x1 gal
- Storage and handling:Room Temperature
- Vapor pressure:<0.1 mm Hg at 20°C
- Description:EcoLite(+)™
- Boiling point:300 °C
- Density (at 20 °C):0.9 - 1.0 g/mL
- Cat. no.:IC-88247504
- Supplier No.:88247504
Specifications
About this item
EcoLite(+)™ is a unique formulation of environmentally safe, nontoxic, and biodegradable media possessing exceptional water and salt holding characteristics.
- Environmentally Safe
- Biodegradable
- Non-Toxic and Non-Flammable
- Ready-To-Use
- Versatile
- Excellent Stability for RIA and Receptor Assays
- Forms No Gel or Phasing
Difficult-to-count RIA samples, basic samples, phosphate buffer samples, and biological fluids can be counted using EcoLite(+)™, with maximum solubilization resulting in
stable counting solutions. Tritium and C counting efficiency of these samples is exceptionally high.
EcoLite(+)™ is a new generation environmentally safe liquid scintillation cocktail that can be used for virtually all aqueous and non-aqueous samples. It is particularly useful for HPLC fractions, urea, phosphate buffers, protein digests, urine and many other applications where conventional cocktails fail to provide homogeneous counting solutions.
EcoLite(+)™ remains monophasic from 0-30% load capacity. Samples are counted by simply adding them to 10-15 mL of EcoLite(+)™, shaking until clear and counting. NOTE: Temperature and salt concentration will affect sample solubility, as well as, the physical characteristics of a surfactant based cocktail. It is recommended that the counting temperature be considered when determining the proper sample to cocktail ratio. Where phase separation occurs, increase the amount of EcoLite(+)™ or dilute with small amounts of distilled water until the sample clears and remains at the desired counting temperature.
Special Characteristics - EcoLite(+)™ is extremely efficient with both H and C samples. It is very fluid, easily pipetted and mixed without sample up to the maximum load capacity. EcoLite(+)™ does not diffuse into or through polyethylene vials, thereby eliminating erroneous external standard quench measurements due to the vial
wall diffusion effect. It has an extremely low vapor pressure which minimizes volatiles and odors under normal laboratory use. It works well with sodium hydroxide samples up to 0.5 N.