To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Chemical method:Stannous chloride, remove molybdenum blue
- Description:Phosphate, ortho CHEMets Visual Kit
- Cat. no.:CXK-8510
- Range:0 - 1 and 1 - 10 ppm
- Supplier No.:K-8510
Specifications
About this item
Ideal for low- to medium-range colorimetric analysis of water quality
CHEMets® test kits utilize self-filling ampules and vacuum pulls the sample up from a snap cup into the ampule when the tip is broken off. After tilting the ampule several times to mix the sample and reagent and waiting for the color to develop (usually 2 minutes or less), the analyst quantifies the result using the provided cylindrical comparator (for concentrations ≤1ppm) or flat comparator (for concentrations ≥1ppm).
Phosphorus occurs naturally in rock formations in the earth’s crust, usually as phosphate. High phosphate concentrations in surface waters may indicate fertilizer runoff, domestic waste discharge, or the presence of industrial effluents or detergents. Although phosphates from these sources are usually poly-phosphates or organically bound, all will degrade to ortho or reactive phosphates with time.
Phosphate measurement is used to control scale and corrosion inhibitor levels in boilers and cooling towers. Both methods Vanadomolybdophosphoric Acid and Stannous Chloride measure reactive phosphate, which will give a positive reaction prior to hydrolysis, and is usually termed ortho-phosphate.