To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Chemical method:Periodic acid / purpald
- Description:Glycol CHEMets Visual Kit
- Cat. no.:CXK-4815
- Range:1 - 15 & 10 - 300 ppm
- Supplier No.:K-4815
Specifications
About this item
CHEMetrics glycol water test kits are used to monitor potable waters for glycol contamination originating from glycol in cooling systems. They are also used to detect glycol in storm water effluent and airplane deicing operations and to monitor glycol recycling operations.
- Ideal for low- to medium-range colorimetric analysis of water quality
CHEMets® test kits utilize self-filling ampules and vacuum pulls the sample up from a snap cup into the ampule when the tip is broken off. After tilting the ampule several times to mix the sample and reagent and waiting for the color to develop (usually 2 minutes or less), the analyst quantifies the result using the provided cylindrical comparator (for concentrations ≤1ppm) or flat comparator (for concentrations ≥1ppm).