Order Entry
ContactUsLinkComponent
CARB DIOX TEST KIT DISS 10-100
Carbon Dioxide (Dissolved), Titrets Titration Cells, CHEMetrics
Catalog #: CXK-1910
Supplier:  CHEMetrics
Small Business Enterprise
CAS Number:  
CARB DIOX TEST KIT DISS 10-100
Carbon Dioxide (Dissolved), Titrets Titration Cells, CHEMetrics
Catalog #: CXK-1910
Supplier:  CHEMetrics
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Chemical method:
    Caustic titrant with pH Indicator
  • Description:
    Carbon Dioxide Titrets Test Kit
  • Cat. no.:
    CXK-1910
  • Increments:
    10, 11, 12, 13, 14, 15, 16, 18, 20, 25, 30, 35, 40, 50, 70, 100 ppm
  • Range:
    0 - 100 ppm
  • Supplier No.:
    K-1910

Specifications

About this item

Titrets® test kits use reverse titration to quantify concentrations in aqueous samples

  • Dependable, sealed single-use titrants
  • Fast, easy procedure — no need to count drops
  • No burettes or other equipment necessary
  • Perform a titration anywhere

After snapping the ampoule tip, the sample is drawn into the ampoule in small doses (with the Titrettor device included in each kit that precisely controls the sample) (Step 1), until a color change signals that the equivalence point has been reached (Step 2). The titration is stopped at the end point and the ampoule is held upright. The liquid level will correspond to a printed scale on the ampoule's outer surface (Step 3).

Carbon dioxide ends up in surface and ground water as a result of animal respiration, decaying organic matter, and the decomposition of some minerals. Surface waters typically contain less than 10 ppm (mg/L) dissolved CO2, while ground waters, particularly if deep, may contain several hundred ppm (mg/L). Carbon dioxide concentration greatly impacts pH. In unpolluted waters, carbon dioxide is a major source of acidity.

Ordering information: Kit comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules with valve assemblies, Activator Solution, titrettor, 25 ml sample cup, and instructions.
Customers Who Bought This Also Bought