Water, OmniSolv®, gradient grade, Supelco®
Catalog #: CAWX0004-1
Supplier: MilliporeSigma
CAS Number: 7732-18-5
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle
- Size:4 L
- Cat. no.:CAWX0004-1
- Supplier No.:WX0004-1
Specifications
Specification Test Results
- Molar Mass:18.02
- Expiration Date:12 months from manufacture date
- Color (APHA):10 max
- Filtered Through 0.2 µm Filter:To pass test
- Fluorescence (as Quinine Base):100 ppt max
- Gradient at 210 nm:0.002 AU max
- Gradient at 254 nm:0.0005 AU max
- Odor:Odorless
- Residue After Evaporation:1 ppm max
- Specific Conductance:2 µmho/cm max
- Total Purgeable Organics:5 ppb max
- UV Abs. at 190 nm:0.010 AU max
- UV Abs. at 200 nm:0.010 AU max
- UV Abs. at 210 nm:0.010 AU max
- UV Abs. at 220 nm:0.010 AU max
- UV Abs. at 230 nm:0.010 AU max
- UV Abs. at 240 nm:0.010 AU max
- UV Abs. at 250 nm:0.005 AU max
- UV Abs. at 280 nm:0.005 AU max
- Supelco®, Water, OmniSolv®, gradient grade:
Specification Test Results
About this item
Synonyms: Hydrogen oxide
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt: 100 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 0 °C
:Flash Pt:
:Flash Pt:
:M.W. 18.02 g/mol
Merck Index:12,10175
:Water
Storage Temperature:Ambient
EINECS:231-791-2
Customers Who Bought This Also Bought