2-Propanol ≥99.8% for HPLC, Supelco®
Catalog #: CAPX1838P-6
Supplier: MilliporeSigma
CAS Number: 67-63-0
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, pre-packed
- Size:1 L
- Cat. no.:CAPX1838P-6
- Supplier No.:PX1838P-6
Specifications
Specification Test Results
- Assay (GC):99.8% min
- Appearance:Clear liquid, free from particulates
- Carbonyl compounds (acetone or propionaldehyde):0.002% max
- Color (APHA):10 max
- Filtered through 0.2 µm filter:To pass test
- Infrared spectrum:Conforms to standard
- Residue after evaporation:5ppm max
- Solubility in water:To pass test
- Titrable acid:0.0001meq/g max
- Titrable base:0.0001meq/g max
- UV Abs. at 210 nm:1.00AU max
- UV Abs. at 220 nm:0.40AU max
- UV Abs. at 230 nm:0.20AU max
- UV Abs. at 245 nm:0.08AU max
- UV Abs. at 260 nm:0.04AU max
- UV Abs. at 275 nm:0.03AU max
- UV Abs. at 300 nm:0.02AU max
- UV Abs. at 350 nm:0.01AU max
- UV Cut-off:210nm max
- Water (H2O):0.10% max
Specification Test Results
About this item
Synonyms: Isopropyl rubbing alcohol,Isopropanol,2-Hydroxy propane,IPA,Dimethyl carbinol,Isopropyl alcohol
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.786 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,8
Flash Pt:12 °C
:Boiling Pt:
:Boiling Pt: 82 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -89 °C|Melting Pt: –89 °C
:Flash Pt:
:Flash Pt: 12 °C
:M.W. 60.1 g/mol
Merck Index:13,05228
:2-Propanol
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
REACH:01-2119457558-25
:3
:II
:GHS02
:GHS07
Customers Who Bought This Also Bought