Methanol ≥99.8% ACS for HPLC, Supelco®
Catalog #: CAMX0475P-1
Supplier: MilliporeSigma
CAS Number: 67-56-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, pre-packed
- Size:4 L
- Cat. no.:CAMX0475P-1
- Supplier No.:MX0475P-1
Specifications
Specification Test Results
- Assay (GC):99.99%
- Carbonyl Compounds (Acetone, Formaldehyde, Acetaldehyde):0.001%
- Color (APHA):10
- Filtered Through 0.2 μm Filter:Passes Test
- Gradient at 254 nm:0.00117 AU
- Infrared Spectrum:Conforms
- Residue after Evaporation:0.5 ppm
- Solubility in Water:Passes Test
- Substances Darkened by Sulfuric Acid:Passes Test
- Substances Reducing Permanganate:Passes Test
- Titrable Acid:0.00027 meq/g
- Titrable Base:0.00009 meq/g
- UV Abs. at 205 nm:0.7563 AU
- UV Abs. at 210 nm:0.3977 AU
- UV Abs. at 220 nm:0.1808 AU
- UV Abs. at 230 nm:0.0844 AU
- UV Abs. at 240 nm:0.0345 AU
- UV Abs. at 260 nm:0.0082 AU
- UV Abs. at 280 nm:0.0051 AU
- UV Abs. at 300 nm:0.0013 AU
- UV Abs. at 350 nm:0.001 AU
- UV Cut-off:203.29 nm
- Water (H₂O):0.0019%
- Supelco®, Methanol ≥99.8% ACS for HPLC:
Specification Test Results
About this item
Synonyms: Hydroxy methane,Carbinol,Wood alcohol
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,8
Flash Pt:11 °C (closed cup)
:Boiling Pt:
:Boiling Pt: 64.6 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -98 °C|Melting Pt: –98 °C
:Flash Pt:
:Flash Pt: 11 °C (closed cup)
:M.W. 32.04 g/mol
Merck Index:13,05984
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
REACH:01-2119433307-44
:3
:II
:11 °C
:GHS02
:GHS06
:GHS08
Customers Who Bought This Also Bought