Acetonitrile ≥99.9%, BAKER ANALYZED® LC-MS for HPLC, for LC-MS, for UV spectrophotometry, J.T.Baker®
Catalog #: CAJT9829-3
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 75-05-8
MOC: Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, amber, narrow mouth
- Size:4 L
- Supplier No.:9829-03
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Assay (CH₃CN):≥99.9 %
- Appearance:Passes Test
- Gradient Elution Test (a.u.) - 254 nm:≤0.0005
- Gradient Elution Test (a.u.) - 210 nm:≤0.002
- Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:≤0.3 ppb
- Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:≤1.0 ppb
- Color (APHA):≤10
- Residue after Evaporation:≤0.8 ppm
- Water (H₂O):≤100 ppm
- LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine):≤50 ppb
- Trace Impurities - Aluminum (Al):≤50.0 ppb
- Trace Impurities - * Calcium (Ca):≤50.0 ppb
- Trace Impurities - Iron (Fe):≤50.0 ppb
- Trace Impurities - Magnesium (Mg):≤50.0 ppb
- Trace Impurities - Lithium (Li):≤30.0 ppb
- Trace Impurities - Nickel (Ni):≤30.0 ppb
- Trace Impurities - Potassium (K):≤50 ppb
- Trace Impurities - * Sodium (Na):≤50.0 ppb
- * May change over time due to extraction from glass container.:
- Filtered through a 0.2 micron filter.:
- J.T.Baker®, Acetonitrile ≥99.9%, BAKER ANALYZED® LC-MS for HPLC, for LC-MS, for UV spectrophotometry:
Specification Test Results
About this item
Synonyms: Methyl cyanide,Ethanoic acid nitrile,Cyanomethane
:Acetonitrile
Boiling Pt:81.6 °C (1013 hPa)
Melting Pt:–45.7 °C
Density:0.782 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,9
:Management of Change (MOC) category = N
Flash Pt:2 °C (closed cup)
:Boiling Pt:
:Boiling Pt: 81.6 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –45.7 °C
:Flash Pt:
:Flash Pt: 2 °C (closed cup)
:M.W. 41.05 g/mol
Merck Index:13,00071
Storage Temperature:Ambient
EINECS:200-835-2
:81.6
UN:1648
REACH:01-2119471307-38
:3
:II
:2 °C
:GHS02
:GHS07
:M.O.C. Category N
Customers Who Bought This Also Bought