Benzyl alcohol 98.0-100.5% NF, Multi-Compendial, J.T.Baker®
Catalog #: CAJT9421-3
Supplier: VWR International
CAS Number: 100-51-6
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, amber, narrow mouth
- Size:4 L
- Supplier No.:9421-03
Specifications
Specification Test Results
- Meets B.P. Chemical Specifications:
- Meets E.P. Chemical Specifications:
- Meets N.F. Requirements:
- Meets J.P. Chemical Specifications:
- GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- NF - Assay (C₆H₅CH₂OH):98.0 - 100.5 %
- NF - Clarity of Solution:Passes Test
- NF - Color of Solution:Passes Test
- NF - Identification:Passes Test
- NF - Peroxide Value:≤5
- NF - Benzaldehyde and Other Related Substances - Benzaldehyde:≤0.15 %
- NF - Related Substances - Cyclohexylmethanol:≤0.10 %
- of Benzyl Alcohol:≤0.04 %
- NF - Benzaldehyde and Other Related Substances - Total Other Peaks <RT:≤0.04 %
- NF - Benzaldehyde and Other Related Substances - Total Other Peaks:≤0.3 %
- Greater Than RT of Benzyl Alcohol:≤0.3 %
- NF - Refractive Index, n²ºD:1.538 - 1.541
- NF - Acidity:Passes Test
- NF - Residue on Evaporation:≤0.05 %
- NF - Benzene:≤2 ppm
- Solubility:Passes Test
- EP/BP - Assay (C₆H₅CH₂OH):98.0 - 100.5 %
- EP/BP - Identification:Passes Test
- EP/BP - Acidity:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Refractive Index, n²º^D:1.538 - 1.541
- EP/BP - Benzaldehyde and Other Related Substances - Benzaldehyde:≤0.15 %
- EP/BP - Related Substances - Cyclohexylmethanol:≤0.10 %
- <RT of Benzyl Alcohol:≤0.04 %
- EP/BP - Benzaldehyde and Other Related Substances - Total Other Peaks:≤0.04 %
- EP/BP - Benzaldehyde and Other Related Substances - TOTAL OTHER PEAKS:≤0.3 %
- >RT OF BENZYL ALCOHOL:≤0.3 %
- EP/BP - Peroxide Value:≤5
- EP/BP - Residue on Evaporation:≤0.05 %
- JP - Assay (C₆H₅CH₂OH):98.0 - 100.5 %
- JP - Identification:Passes Test
- JP - Refractive Index, n²º^D:1.538 - 1.541
- JP - Specific Gravity at 20°/20°C:1.043 - 1.049
- JP - Clarity and Color of Solution:Passes Test
- JP - Acidity:Passes Test
- JP - Benzaldehyde and Other Related Substances - Benzaldehyde:≤0.15 %
- JP - Related Substances - Cyclohexylmethanol:≤0.10 %
- JP - Benzaldehyde and Other Related Substances - Total Other Peaks <RT:≤0.04 %
- OF BENZYL ALCOHOL:≤0.3 %
- JP - Benzaldehyde and Other Related Substances - TOTAL OTHER PEAKS >RT:≤0.3 %
- JP - Peroxide Value:≤5
- JP - Residue on Evaporation:≤0.05 %
- This product is not intended for use in preparing parenteral dosage:
- forms.:
- The Class 1 solvent Benzene is likely to be present below 2 ppm. The:
- actual analysis of Benzene is reported for each lot.:
- The Class 2 solvent Toluene is likely to be present and is process:
- controlled to <30 ppm.:
- No other Class 1, 2 or 3 residual solvents are likely to be present.:
Specification Test Results
About this item
Synonyms: Phenylmethanol
Boiling Pt:205 °C (1013 hPa)
Melting Pt:–15.3 °C
Density:1.05 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,0-100,5
:Management of Change (MOC) category = R
Flash Pt:101 °C
:Boiling Pt:
:Boiling Pt: 205 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –15.3 °C
:Flash Pt:
:Flash Pt: 101 °C
:M.W. 108.14 g/mol
Merck Index:13,01126
Storage Temperature:Ambient
EINECS:202-859-9
:GHS07
:M.O.C. Category R
Customers Who Bought This Also Bought