n-Hexane ≥95.0% (by GC, corrected for water content) for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Catalog #: CAJT9304-3
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 110-54-3
MOC: Non-Regulated, With InformationCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, amber, narrow mouth
- Size:4 L
- Cat. no.:CAJT9304-3
- Supplier No.:9304-03
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm:≤0.005
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm:≤0.005
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm:≤0.008
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm:≤0.08
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm:≤0.2
- Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:≤192
- Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:≤0.3 ppb
- Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:≤1.0 ppb
- Assay (Total Saturated C₆ Isomers) (by GC, corrected for water):≥99.5 %
- Assay (as n-Hexane) (by GC, corrected for water):≥95.0 %
- Residue after Evaporation:≤2.0 ppm
- Substances Darkened by H₂SO₄:Passes Test
- Water (by KF, coulometric):≤0.01 %
- Filtered through a 0.2 micron filter.:
- Packaged under Nitrogen.:
- J.T.Baker®, n-Hexane ≥95.0% (by GC, corrected for water content) for HPLC, UHPLC, for spectrophotometry:
Specification Test Results
About this item
Synonyms: Hexyl hydride,Hexanen
:n-Hexane
Boiling Pt:69 °C (1013 hPa)
Melting Pt:–94.3 °C
Density:0.659 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95,0
:Management of Change (MOC) category = NI
Flash Pt:–22 °C
:Boiling Pt:
:Boiling Pt: 69 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –94.3 °C
:Flash Pt:
:Flash Pt: –22 °C
:M.W. 86.18 g/mol
Merck Index:13,04712
Storage Temperature:Ambient
EINECS:203-777-6
:69
UN:1208
:3
:II
:–22 °C
:GHS02
:GHS08
:GHS07
:GHS09
:M.O.C. Category NI
Customers Who Bought This Also Bought