Sodium hydroxide 10 N in aqueous solution, Biotech Reagent, J.T.Baker®
Catalog #: CAJT5000-02
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 1310-73-2
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:PE bottle, narrow mouth
- Size:1 L
- Supplier No.:5000-02
Specifications
Specification Test Results
- GMP Manufactured Product:
- IMPORTANT: Material will freeze if stored below 41 °F (5°C).:
- Protect from air to avoid absorption of carbon dioxide.:
- CAUTION: For Manufacturing, processing or repackaging:
- Appearance (Clear, colorless liquid):Passes Test
- Normality:9.9 - 10.1
- Sodium Carbonate (Na₂CO₃):≤0.6 %
- Chloride (Cl):≤20 ppm
- ACS - Heavy Metals (as Pb):≤1 ppm
- Trace Impurities - Iron (Fe):≤2 ppm
- Endotoxin Concentration (EU/mL):≤2.0
- Made from U.S.P. Purified Water and Sodium Hydroxide pellets, N.F. (Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical Specifications:
- No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:
- IMPORTANT: Material will freeze if stored below 41°F (5°C).:
- Filtered through a 0.2 micron filter.:
- Sodium hydroxide 10 N in aqueous solution, Biotech Reagent, J.T.Baker®:
Specification Test Results
About this item
Synonyms: Caustic soda
Boiling Pt:119…135 °C (1013 hPa)
Melting Pt:9…10 °C
Density:1.32 g/cm³ (20 °C)
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 119…135 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 323 °C|Melting Pt: 9…10 °C
:Flash Pt:
:Flash Pt:
:M.W. 40 g/mol
EINECS:215-185-5
:119…135 °C (1013 hPa)
UN:1824
:8
:II
:GHS05
:GHS07
Storage Temperature:Ambient
:M.O.C. Category R
:12,08772
:14,08627
Customers Who Bought This Also Bought