Sodium dodecyl sulfate (SDS) ≥99%, Ultrapure Bioreagent, J.T.Baker®
Catalog #: CAJT4095-4
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 151-21-3
MOC: Non-RegulatedCertificates
Specifications
- Packaging:Poly bottle, white, blue T/E cap
- Size:100 g
- Supplier No.:4095-04
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Appearance:Passes Test
- Assay (as C₁₂H₂₅OSO₃Na):≥99 %
- Absorbance of a 3% w/v Aqueous Solution at 280 nm (1-cm path):≤0.1
- pH of 1% Solution at 25°C:5.0 - 8.0
- Chloride (Cl):≤0.03 %
- Nitrogen (N):≤5 ppm
- Phosphate (PO₄):≤1 ppm
- Heavy Metals (as Pb):≤2 ppm
- Trace Impurities - Iron (Fe):≤1 ppm
- DNase Activity (None Detected):Passes Test
- RNase Activity:Passes Test
- J.T.Baker®, Sodium dodecyl sulfate (SDS) ≥99%, Ultrapure Bioreagent:
Specification Test Results
About this item
Synonyms: Sodium lauryl sulfate,Dodecyl sulfate sodium salt,Lauryl sulfate sodium salt,Dodecyl sulfuric acid sodium salt,Dodecyl sulphate sodium salt,Sodium dodecyl sulfate,Dodecyl sulphuric acid sodium salt,Lauryl sulphate sodium salt
:Sodium dodecyl sulfate (SDS)
Melting Pt:204…207 °C
Density:0.40 g/cm³
ADR:4.1 (6.1),II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99
:Management of Change (MOC) category = N
Flash Pt:>100 °C
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 204 to 207 °C|Melting Pt: 204…207 °C
:Flash Pt:
:Flash Pt: >100 °C
:M.W. 288.38 g/mol
Merck Index:13,08710
Storage Temperature:Ambient
EINECS:205-788-1
UN:2926
:4.1 (6.1)
:II
:>100 °C
:GHS02
:GHS06
:GHS09
:M.O.C. Category N
Customers Who Bought This Also Bought