Sodium chloride 99.0-100.5% (dried basis), granular USP, Multi-Compendial, J.T.Baker®
Catalog #: CAJT3627-1
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 7647-14-5
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, amber, wide mouth, black T/E cap
- Size:500 g
- Supplier No.:3627-01
Specifications
Specification Test Results
- Meets J.P. Chemical Specifications:
- Meets U.S.P Requirements:
- Meets B.P. Chemical Specifications:
- Meets E.P. Chemical Specifications:
- GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Acidity or Alkalinity:Passes Test
- USP - Appearance of Solution:Passes Test
- USP - Aluminum (Al):≤0.2 ppm
- USP - Arsenic (As):≤1 ppm
- USP - Barium (Ba):Passes Test
- USP - Bromide (Br):≤100 ppm
- USP - Ferrocyanide:Passes Test
- USP - Iodides (I):Passes Test
- USP - Iron (Fe):≤2 ppm
- USP - Loss on Drying at 105°C:≤0.5 %
- USP - Magnesium and Alkaline Earth Metals (as Ca):≤100 ppm
- USP - Nitrite (NO₂) (Absorbance Value):≤0.01
- USP - Phosphate (PO₄):≤25 ppm
- USP - Potassium (K):≤500 ppm
- USP - Sulfate (SO₄):≤200 ppm
- USP - Assay (NaCl) (dried basis):99.0 - 100.5 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Aluminum (Al):≤0.2 ppm
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Acidity or Alkalinity:Passes Test
- EP/BP - Bromide (Br):≤100 ppm
- EP/BP - Ferrocyanide:Passes Test
- EP/BP - Iodide (I):Passes Test
- EP/BP - Nitrite (NO₂):≤0.01
- EP/BP - Phosphate (PO₄):≤25 ppm
- EP/BP - Sulfate (SO₄):≤200 ppm
- EP/BP - Arsenic (As):≤1 ppm
- EP/BP - Barium (Ba):Passes Test
- EP/BP - Iron (Fe):≤2 ppm
- EP/BP - Magnesium and Alkaline Earth Metals:≤100 ppm
- EP/BP - Loss on Drying at 105°C:≤0.5 %
- EP/BP - Potassium (K):≤500 ppm
- EP/BP - Assay (NaCl) (dried basis):99.0 - 100.5 %
- EP/BP - Endotoxin Concentration, IU/g:≤2.5
- JP - Assay (NaCl) (dried basis):99.0 - 100.5 %
- JP - Identification A:Passes Test
- JP - Identification B:Passes Test
- JP - Acidity or Alkalinity:Passes Test
- JP - Clarity and Color of Solution:Passes Test
- JP - Bromide (Br):Passes Test
- JP - Ferrocyanide:Passes Test
- JP - Iodide (I):Passes Test
- JP - Iron (Fe):Passes Test
- JP - Magnesium and Alkaline Earth Metals:Passes Test
- JP - Phosphate (PO₄):Passes Test
- JP - Sulfates (SO₄):Passes Test
- JP - Heavy Metals (as Pb):≤3 ppm
- JP - Barium (Ba):Passes Test
- JP - Arsenic (As):≤2 ppm
- JP - Loss on Drying:≤0.5 %
- Total Aerobic Microbial Count, For Information Only:
- Total Yeasts and Molds Count, For Information Only:
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Must be subjected to further processing during the preparation of:
- injectable dosage forms:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- J.T.Baker®, Sodium chloride 99.0-100.5% (dried basis), granular USP, Multi-Compendial:
Specification Test Results
About this item
Synonyms: Saline,Sea salt
:Sodium chloride
Boiling Pt:1413 °C (1013 hPa)
Melting Pt:801 °C
Density:2.16 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-100,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 1413 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 801 °C
:Flash Pt:
:Flash Pt:
:M.W. 58.44 g/mol
Merck Index:14,08599
Storage Temperature:Ambient
EINECS:231-598-3
:GHS07
:M.O.C. Category R
Customers Who Bought This Also Bought