Lactic acid 85%, BAKER ANALYZED® ACS, J.T.Baker®
Catalog #: CAJT0194-1
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 50-21-5
MOC: Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, amber, narrow mouth
- Size:500 ml
- Cat. no.:CAJT0194-1
- Supplier No.:0194-01
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay (as CH₃CHOHCOOH):85.0 - 90.0 %
- ACS - Residue after Ignition:≤ 0.02 %
- ACS - Chloride (Cl):≤ 0.001 %
- ACS - Sulfate (SO₄):≤ 0.002 %
- ACS - Substances Darkened by H₂SO₄:Passes Test
- ACS - Heavy Metals (by ICP-OES):≤ 5 ppm
- Trace Impurities - Iron (Fe):≤ 5 ppm
- The reagent generally available is a mixture of lactic acid (CH₃CHOHCOOH) and lactic acid lactate (C₆H₁₀O₅).:
- J.T.Baker®, Lactic acid 85%, BAKER ANALYZED® ACS:
Specification Test Results
About this item
Synonyms: DL-Lactic acid,2-Hydroxypropionic acid
Boiling Pt:122 °C (20 hPa)
Density:1.20 g/cm³
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:85%
:85,0-90,0
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt: 122 °C (20 hPa)
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 90.08 g/mol
Storage Temperature:Ambient
EINECS:200-018-0
UN:3265
:8
:II
:GHS05
:M.O.C. Category N
Customers Who Bought This Also Bought