n-Hexane ≥95%, OmniSolv®, Supelco®
Catalog #: CAHX0295-1
Supplier: MilliporeSigma
CAS Number: 110-54-3
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle
- Size:4 L
- Cat. no.:CAHX0295-1
- Supplier No.:HX0295-1
Specifications
Specification Test Results
- Assay (n-hexane):95% min
- Boiling range:66.1-69.4°C
- Capillary ECD responsive substances (as PCNB):5ng/L max
- Capillary FID responsive substances (as decane):3µg/L max
- Color (APHA):10 max
- Filtered through 0.2 µm filter:To pass test
- Fluorescence (as quinine base):250ppt max
- Form:Clear liquid
- Infrared spectrum:Conforms to standard
- Residue after evaporation:1ppm max
- UV Abs. at 195 nm:1.00AU max
- UV Abs. at 200 nm:0.50AU max
- UV Abs. at 210 nm:0.20AU max
- UV Abs. at 220 nm:0.05AU max
- UV Abs. at 230 nm:0.05AU max
- UV Abs. at 250 nm:0.01AU max
- UV Abs. at 280 nm:0.005AU max
- UV Cut-off:195nm max
- Water (H2O):0.01% max
Specification Test Results
About this item
Synonyms: Hexyl hydride,Hexanen
:n-Hexane
Boiling Pt:69 °C (1013 hPa)
Melting Pt:–94.3 °C
Density:0.659 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95
Flash Pt:–22 °C
:Boiling Pt:
:Boiling Pt: 69 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -94.3 °C|Melting Pt: –94.3 °C
:Flash Pt:
:Flash Pt: –22 °C
:M.W. 86.18 g/mol
Merck Index:13,04712
EINECS:203-777-6
UN:1208
:3
:II
:–22 °C
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Customers Who Bought This Also Bought