N,N-Dimethylformamide ≥99.9%, OmniSolv®, Supelco®
Catalog #: CADX1726-1
Supplier: MilliporeSigma
CAS Number: 68-12-2
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle
- Size:4 L
- Cat. no.:CADX1726-1
- Supplier No.:DX1726-1
Specifications
Specification Test Results
- Assay (GC):99.9% min
- Color (APHA):10 max
- Expiration date:24 months
- Filtered through 0.2 µm filter:To pass test
- Form:Clear liquid
- Infrared spectrum:Conforms to standard
- Residue after evaporation:5ppm max
- UV Abs. at 268 nm:1.00AU max
- UV Abs. at 270 nm:0.75AU max
- UV Abs. at 275 nm:0.30AU max
- UV Abs. at 295 nm:0.07AU max
- UV Abs. at 310 nm:0.03AU max
- UV Abs. at 340 nm:0.01AU max
- UV Cut-off:268nm max
- Water (H2O):0.05% max
Specification Test Results
About this item
Synonyms: Formic acid dimethylamide,DMFA,Formyldimethylamine,N-Formyldimethylamine,Dimethylformamide,NSC 5356
Boiling Pt:153 °C (1013 hPa)
Melting Pt:–61 °C
Density:0.949 g/cm³ (20 °C)
ADR:3,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,9
Flash Pt:57.5 °C (closed cup)
:Boiling Pt:
:Boiling Pt: 153 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -61 °C|Melting Pt: –61 °C
:Flash Pt:
:Flash Pt: 57.5 °C (closed cup)
:M.W. 73.09 g/mol
Merck Index:13,03269
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
REACH:01-2119475605-32
:3
:III
:57.5 °C
:GHS02
:GHS08
:GHS07
Customers Who Bought This Also Bought