Dichloromethane ≥99.9% stabilized, OmniSolv®, Supelco®
Catalog #: CADX0831-1
Supplier: MilliporeSigma
CAS Number: 75-09-2
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle
- Size:4 L
- Cat. no.:CADX0831-1
- Supplier No.:DX0831-1
Specifications
Specification Test Results
- Assay (GC):99.9% min
- Capillary ECD responsive substances (as PCNB):2ng/L max
- Capillary FID responsive substances (as decane):3µg/L max
- Color (APHA):10 max
- Filtered through 0.2 µm filter:To pass test
- Fluorescence (as quinine base):500ppt max
- Form:Clear liquid
- Free halogens:To pass test
- Identity (IR-spectrum):Conforms
- Residue after evaporation:1ppm max
- Titrable acid:0.2µeq/g max
- UV Abs. at 231 nm:1.00AU max
- UV Abs. at 235 nm:0.40AU max
- UV Abs. at 240 nm:0.20AU max
- UV Abs. at 250 nm:0.01AU max
- UV Abs. at 260 nm:0.005AU max
- UV Cut-off:231nm max
- Water (H2O):0.005% max
Specification Test Results
About this item
Synonyms: Methylene chloride
Boiling Pt:39.8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1.322 g/cm³ (20 °C)
ADR:6.1,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,9
:Boiling Pt:
:Boiling Pt: 39.8 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -95 °C|Melting Pt: –95 °C
:Flash Pt:
:Flash Pt:
:M.W. 84.93 g/mol
Merck Index:12,06140
:Dichloromethane
Storage Temperature:Ambient
EINECS:200-838-9
UN:1593
:6.1
:III
:GHS08
:GHS07
:stab
Customers Who Bought This Also Bought