Order Entry
ContactUsLinkComponent
COD TEST KIT 0-15000PPM 100VL
COD Vials Kits, Chemetrics
Catalog #: CACXK7375
Supplier:  Chemetrics
CAS Number:  
COD Vials Kits, Chemetrics
Catalog #: CACXK7375
Supplier:  Chemetrics
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Chemical method:
    Dichromate reactor digestion
  • Description:
    COD Vial Kit, 98 vials
  • Cat. no.:
    CACXK7375
  • Range:
    0 - 15000 ppm
  • Supplier No.:
    K-7375
  • No. of Tests:
    97

Specifications

About this item

The kit provides two dichromate reactor digestion methods such as USEPA-accepted method, and a Mercury-free method for fast, easy, safe determinations of low, mid, and high-range COD levels in wastewater.

  • Leakproof reagent vials
  • Fast safe and easy COD determination

A fresh reagent ampoule blank must be prepared for each series of tests, therefore the number of samples that can be tested with each kit will vary.

Delivery: Kits come in a cardboard box and contain everything needed to perform the tests (except distilled water) and instruction book.
Caution: These kits require the use of a Digestor Block along with a CHEMetrics Photometer, a COD Photometer, or a spectrophotometer capable of accepting a 16 mm round cell. All these instruments are sold separately.
Unless indicated as mercury-free, the kits contain mercury. Dispose according to local, state or federal laws.
Customers Who Bought This Also Bought