Order Entry
ContactUsLinkComponent
Fibronectin
Corning® Human Fibronectin
Catalog #: CACB356008
Supplier:  Corning
CAS Number:  
Fibronectin
Corning® Human Fibronectin
Catalog #: CACB356008
Supplier:  Corning
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Species:
    Human
  • Size:
    5 mg
  • Protein/peptide name:
    Fibronectin
  • Cat. no.:
    CACB356008
  • Supplier No.:
    356008

Specifications

About this item

Fibronectin is found in interstitial matrix and plasma. The principal function of fibronectin appears to be in cellular migration during wound healing and development. It can be used as a thin coating on tissue culture surfaces to promote attachment, spreading and proliferation of a variety of cell types.

Quality controlled by SDS-PAGE and tested and found negative for bacteria, fungi and mycoplasma. Source material tested for hepatitis B antigen and HIV-1 antibody.

Customers Who Bought This Also Bought