Order Entry
ContactUsLinkComponent
Endothelial Cell Growth Supplement (ECGS)
Corning® Endothelial Cell Growth Supplement, Corning
Catalog #: CACB356006
Supplier:  Corning
CAS Number:  
Corning® Endothelial Cell Growth Supplement, Corning
Catalog #: CACB356006
Supplier:  Corning
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Regulatory status:
    RUO
  • Cat. no.:
    CACB356006
  • Source:
    Bovine brain
  • Shelf Life:
    790 days
  • Volume:
    100 mg
  • Supplier No.:
    356006

Specifications

About this item

Endothelial Cell Growth Supplement (ECGS) is a broadly used supplement to culture a variety of cells, particularly endothelial cells.

ECGS contains various growth factors (e.g., acidic FGF or ECCF-a).

Tested for ability to promote proliferation of Fetal Bovine Heart Endothelial Cells (FBHEC) in medium containing 10% newborn calf serum. Filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi, and mycoplasma.

Customers Who Bought This Also Bought