Order Entry
ContactUsLinkComponent
MITO™+ Serum Extender
Corning® MITO+ Serum Extender, Corning
Catalog #: CACB355006
Supplier:  Corning
CAS Number:  
Corning® MITO+ Serum Extender, Corning
Catalog #: CACB355006
Supplier:  Corning
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Regulatory status:
    RUO
  • Shelf Life:
    790 days
  • Volume:
    5 ml
  • Supplier No.:
    355006

Specifications

About this item

MITO+ Serum Extender is a concentrated, fully defined formulation of hormones, growth factors (EGF and FGF), and other metabolites (insulin and steriod hormones)

It can be used to culture a variety of cells under serum-free or serum-reduced conditions.

Tested for ability to promote proliferation of BALB/c-3T3 cells in serum-reduced medium. Filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi and mycoplasma.

(5ml; 5L equivalent).

Customers Who Bought This Also Bought