Order Entry
ContactUsLinkComponent
ITS+ Premix
Corning® ITS Universal Culture Supplement Premix, Corning
Catalog #: CACB354352
Supplier:  Corning
CAS Number:  
Corning® ITS Universal Culture Supplement Premix, Corning
Catalog #: CACB354352
Supplier:  Corning
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    ITS+ Universal Culture Supplement Premix
  • Regulatory status:
    RUO
  • Shelf Life:
    790 days
  • Volume:
    20 ml
  • Supplier No.:
    354352

Specifications

About this item

ITS contains insulin, human transferrin, and selenous acid, the three most universally essential components of defined culture media.

They stimulate cell proliferation of a variety of cells under serum-reduced conditions.

Tested for ability to promote proliferation of HeLa cells in serum-free medium. Filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi and mycoplasma.

Customers Who Bought This Also Bought