Order Entry
ContactUsLinkComponent
Cell Recovery Solution
Corning® Cell Recovery Solution, Corning
Catalog #: CACB354253
Supplier:  Corning
CAS Number:  
Cell Recovery Solution
Corning® Cell Recovery Solution, Corning
Catalog #: CACB354253
Supplier:  Corning
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Cell Recovery Solution
  • Size:
    100 ml
  • Storage Temperature:
    +2…+8 °C
  • Supplier No.:
    354253

Specifications

About this item

Corning® Cell Recovery Solution allows for the recovery of cells cultured on Corning® Matrigel® Basement Membrane Matrix for subsequent biochemical analysis. Corning® Cell Recovery Solution depolymerises Corning® Matrigel® Matrix gels without enzymatic digests and lengthy incubation periods at high temperatures. Cells are released without damage thereby avoiding biochemical changes during incubation and digestion of extracellular portions of cell-surface receptors and adhesion molecules.

  • Non-enzymatic proprietary solution
  • Tested for ability to depolymerise a 1 mm thick layer of gelled BD Matrigel™ matrix after one hour at 2 to 8 °C
  • Filtered and tested and found negative for bacteria, fungi, and mycoplasma
  • Stable for at least three months at 2 to 8 °C
Customers Who Bought This Also Bought