Order Entry
ContactUsLinkComponent
COLLAGEN RATTAIL HI PROT 100MG
Corning® rat collagen I (from tail tendon)
Catalog #: CACB354249
Supplier:  Corning
CAS Number:  
COLLAGEN RATTAIL HI PROT 100MG
Corning® rat collagen I (from tail tendon)
Catalog #: CACB354249
Supplier:  Corning
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Source:
    Tail
  • Species:
    Rat
  • Size:
    100 mg
  • Protein/peptide name:
    Collagen I
  • Cat. no.:
    CACB354249
  • Supplier No.:
    354249

Specifications

About this item

Collagen I is found in most tissues and organs, but is most plentiful in dermis, tendon, and bone. Used as a thin coating in cell and tissue culture, it is often used to enhance cell attachment and proliferation. When applied as a gel it can be used to promote expression of cell-specific morphology and function. Collagen I is commonly used to culture endothelial cells, hepatocytes, muscle cells, and a variety of other cell types. The HC formulation of rat tail collagen I is used for three dimensional applications requiring a sturdy gel that provides a maximal 3D support matrix (guidelines are included with product).

Quality control: All preparations are quality controlled by SDS-PAGE and tested and found negative for bacteria, fungi, and mycoplasma. Source material for human collagen I tested for hepatitis B antigen and HIV-1 antibody.

Customers Who Bought This Also Bought