Order Entry
ContactUsLinkComponent
Matrigel* Basement Membrane Matrix
Corning® Matrigel® Basement Membrane Matrix, Corning
Catalog #: CACB354234
Supplier:  Corning
CAS Number:  
Corning® Matrigel® Basement Membrane Matrix, Corning
Catalog #: CACB354234
Supplier:  Corning
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Matrigel™ Matrix
  • Supplier No.:
    354234

Specifications

About this item

Growth Factor Reduced (GFR) formulation is well suited for applications that require a more highly defined basement membrane preparation

  • Mouse colonies routinely screened for pathogens via Mouse Antibody Production (MAP) testing
  • Extensive PCR testing is performed to screen for a number of pathogens
  • Tested and found negative for bacteria, fungi, and mycoplasma
  • Protein concentrations determined by Lowry method
  • Endotoxin units measured by Limulus Amoebocyte Lysate assay
  • Matrigel Matrix gel stability tested for a period of 14 days at 37°C
  • Biological activity determined for each lot using a neurite outgrowth assay.

Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors.

Application
• Cell Growth and Differentiation
• Metabolism/Toxicology Studies
• Invasion Assays
• In Vitro and In Vivo Angiogenesis Assays
• In Vivo Angiogenesis Studies and Augmentation of Tumors in Immunosuppressed Mice

Preparation and Storage
Store at -20°C. Avoid multiple freeze-thaws

Note: For research use only. Not intended for diagnostic or therapeutic procedures.

When warmed, the solution polymerizes to produce a biologically active matrix material resembling the mammalian cellular basement membrane. It is ideal for use with in vitro cell cultures of a broad range of cell types, including both primary and stem cell applications.

Customers Who Bought This Also Bought