Order Entry
ContactUsLinkComponent
Laminin
Corning® Mouse Laminin
Catalog #: CACB354232
Supplier:  Corning
CAS Number:  
Laminin
Corning® Mouse Laminin
Catalog #: CACB354232
Supplier:  Corning
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Species:
    Mouse
  • Size:
    1 mg
  • Protein/peptide name:
    Laminin
  • Supplier No.:
    354232

Specifications

About this item

Laminin, a major component of basement membranes, has numerous biological activities including promotion of cell adhesion, migration, growth, and differentiation, including neurite outgrowth. It can be used as a thin coating on tissue culture surfaces or as a soluble additive to culture medium. The Laminin/Entactin Complex, high concentration (HC) is a special formulation that has been developed for three-dimensional (3D) culture.

Quality controlled by SDS-PAGE and tested and found negative for bacteria, fungi, and mycoplasma.

Customers Who Bought This Also Bought