ContactUsLinkComponent
HUVEC2 CELLS CRYOVIAL
Corning® Human Umbilical Vein Endothelial Cells, HUVEC-2 Cells, Corning
Catalog #: CACB354151
Supplier:  Corning
CAS Number:  
HUVEC2 CELLS CRYOVIAL
Corning® Human Umbilical Vein Endothelial Cells, HUVEC-2 Cells, Corning
Catalog #: CACB354151
Supplier:  Corning
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Cell Format:
    Cryopreserved
  • Description:
    Human Umbilical Vein Endothelial Cells, 10% DMSO Media
  • Size:
    1 Cryovial/Pack
  • Cat. no.:
    CACB354151
  • Supplier No.:
    354151
Specifications
About this item
HUVEC-2 cells are qualified for use in Corning® BioCoat™angiogenesis systems: endothelial cell migration
The cells may be used in Corning BioCoat endothelial cell invasion and tube formation assays. They are used as model for studying endothelial differentiation and modulation of endothelial tube formation by antiangiogenic agents.
Ordering information: Includes Corning®Human Umbilical Vein Endothelial Cells, HUVEC-2 Cells (>5×105 Cells).
Customers Who Bought This Also Bought