Specifications
- Volume:50 µl
- Cat. no.:CABKA21586
- Filter:+
- Sterile:+
- Tip Style:Biomek® P50 Tips, with barrier
- For Use With:AP96 multichannel heads, Span-8 pod, and Biomek 4000 pipette tools
- Supplier No.:A21586
Specifications
About this item
Biomek® Pipette Tips from Beckman Coulter® are the only tips on the market that are designed, validated, and approved for use on Biomek® Liquid Handlers.
Products that are designated as Sterile are either sterilized using Gamma radiation or ethylene oxide (EtO) gas in accordance with ANSI/AAMI/150 11135 or ANSI/AAMI/ISO 11137 guidelines. The sterilization processes certify a sterility assurance level of 10-6. Biomek® P30, P30XL, P20, P50, and P250 tips are EtO gas-sterilized for critical low-volume pipetting performance.
- Color-coded racks
- Easy visual checks
Biomek® Pipette Tips from Beckman Coulter® are the only tips on the market that are designed, validated, and approved for use on Biomek® Liquid Handlers.
Tip definitions, pipetting techniques, and templates are built into Biomek® software. For best in total system performance, use only Beckman Coulter® validated pipette tips on Biomek® workstations.
Tip definitions, pipetting techniques, and templates are built into Biomek® software. For best in total system performance, use only Beckman Coulter® validated pipette tips on Biomek® workstations.
Products that are designated as Sterile are either sterilized using Gamma radiation or ethylene oxide (EtO) gas in accordance with ANSI/AAMI/150 11135 or ANSI/AAMI/ISO 11137 guidelines. The sterilization processes certify a sterility assurance level of 10-6. Biomek® P30, P30XL, P20, P50, and P250 tips are EtO gas-sterilized for critical low-volume pipetting performance.
Tip racks are color-coded and matched in Biomek® software for easy-to-identify visual checks during method development and confirmation of the deck before a run.