Acetic acid glacial 99.7-100.5% ACS, meets analytical specification of USP, FCC, VWR Chemicals BDH®
Catalog #: CABDH3092500ML
Supplier: BDH
CAS Number: 64-19-7
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Poly bottle
- Size:500 ml
- Supplier No.:3092
Specifications
Specification Test Results
- Assay (CH3COOH):99.7% min.(ACS)/99.5-100.5%(USP/FCC)
- Acetic Anhydride:0.01% max.(ACS)
- Chloride (as Cl):To Pass Test(USP)
- Chloride (Cl):1 ppm max.(ACS)
- Color (APHA):10 max.(ACS)
- Congealing Temperature:15.6 C min.(USP/FCC)
- Dilution Test:To pass test(ACS)
- Heavy Metals (as Pb):0.5 ppm max.(ACS)
- Identification::acetate To Pass Test(USP/FCC)
- Iron (Fe):0.2 ppm max.(ACS)
- Lead (Pb):0.5 mg/kg max.(FCC)
- Nonvolatile residue:1.0mg max.(USP)/0.005% max.(FCC)
- Oxidizable substances:To pass test(USP/FCC)
- Residue After Evaporation:0.001% max.(ACS)
- Substances Reducing Dichromate:To pass test (ACS)
- Substances Reducing Permanganate:To pass test (ACS)
- Sulfate:1 ppm max.(ACS)
- Sulfate (SO4):To Pass Test(USP)
- Titrable Base:0.0004 meq/g max.(ACS)
- Manufacture Date:Record Actual
- Expiration date:Record Actual
- VWR Chemicals BDH®, Acetic acid glacial 99.7-100.5% ACS, meets analytical specification of USP, FCC:
Specification Test Results
About this item
Synonyms: Vinegar acid,Methanecarboxylic acid
Boiling Pt:118 °C (1013 hPa)
Melting Pt:17 °C
Density:1.05 g/cm³ (20 °C)
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,7-100,5
Flash Pt:38.5 °C
:Boiling Pt:
:Boiling Pt: 118 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 17 °C
:Flash Pt:
:Flash Pt: 38.5 °C
:M.W. 60.05 g/mol
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
REACH:01-2119475328-30
:8
:II
:38.5 °C
:GHS02
:GHS05
Customers Who Bought This Also Bought